Learn More
R&R Lotions : PREGLOVE FRAG LOTION 8OZ 24P
SDP
Catalog No. NC0855023
Encompass
•Helps relieve dermatitis symptoms by moisturizing the skin •Static dissipative for use in clean rooms and electrostatic protected areas •Compatible with latex and nitrile gloves to reduce the risk of glove degradation, and with CHG anti-pathogen to reduce the risk of deactivating interference •Does not contain glycerin, mineral oil, silicones, or lanolin, any of which may contaminate integrated circuit boards during manufacturing or handling •8 oz. squeeze bottle for portability
Catalog No. NC0855023 Supplier R&R Lotions Supplier No. ICL8
•Helps relieve dermatitis symptoms by moisturizing the skin •Static dissipative for use in clean rooms and electrostatic protected areas •Compatible with latex and nitrile gloves to reduce the risk of glove degradation, and with CHG anti-pathogen to reduce the risk of deactivating interference •Does not contain glycerin, mineral oil, silicones, or lanolin, any of which may contaminate integrated circuit boards during manufacturing or handling •8 oz. squeeze bottle for portability