Learn More
Lonza Walkersville PrEBM Prostate Epithelial Cell Growth Basal Medium
Catalog No. NC9780053
Encompass
PrEBM is a prostate epithelial cell growth basal medium sold separately. This can be used with 1 kit of CC-4177 (prostate epithelial cell growth medium SingleQuots Supplements and Growth Factors). The full PrEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3166.PrEGM Bulletkit, Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. PrEGM Medium contains no serum. Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9780053 Supplier Lonza Walkersville Supplier No. CC3165
PrEBM is a prostate epithelial cell growth basal medium sold separately. This can be used with 1 kit of CC-4177 (prostate epithelial cell growth medium SingleQuots Supplements and Growth Factors). The full PrEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3166.PrEGM Bulletkit, Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. PrEGM Medium contains no serum. Content:1 x Bottle Basal Medium 500 mL