Learn More
Lonza Walkersville PrEGM Prostate Epithelial Cell Growth Medium BulletKit
Supplier:  Lonza Walkersville CC3166
Encompass
Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. The BulletKit consists of basal medium and supplements packaged together for convenience. PrEGM Medium contains no serum.Content:1 x PrEBM Basal Medium (CC-3165), 500 mL1 x PrEGM SingleQuots Supplement Pack (CC-4177) containing:1 x Orange Cap Vial with BPE, 2.00 mL1 x Amber Vial with Triiodothyronine, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Epinephrine, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Amber Vial with Retinoic Acid, 0.50 mL
Catalog No. NC9462402