Learn More
Lonza Walkersville PrEGM Prostate Epithelial Cell Growth Medium SingleQuots Supplements and Growth Factors
Catalog No. NC9846744
Encompass
PrEGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the PrEGM Prostate Epithelial Cell Growth Medium BulletKit (Catalog Number CC-3166). The full PrEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3166.PrEGM Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. PrEGM Medium contains no serum.Content:1 x Orange Cap Vial with BPE 2.00 mL1 x Amber Vial with Triiodothyronine 0.50 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL1 x Amber Vial with Epinephrine 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Amber Vial with Retinoic Acid 0.50 mL
Catalog No. NC9846744 Supplier Lonza Walkersville Supplier No. CC4177
PrEGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the PrEGM Prostate Epithelial Cell Growth Medium BulletKit (Catalog Number CC-3166). The full PrEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3166.PrEGM Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. PrEGM Medium contains no serum.Content:1 x Orange Cap Vial with BPE 2.00 mL1 x Amber Vial with Triiodothyronine 0.50 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL1 x Amber Vial with Epinephrine 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Amber Vial with Retinoic Acid 0.50 mL