Learn More
Medchemexpress LLC Pregn-4-en-18-al,11,21-dihydroxy-3,20-dioxo-, (11β)-d4 | 99.30% | 364.47 | 100 UG
SDP
Catalog No. 5000436071
Encompass_Preferred
Aldosterone-d4 is a deuterium-labeled version of Aldosterone, which is the primary mineralocorticoid. This steroid hormone is synthesized and secreted in response to the activation of the renin-angiotensin system (RAS) or high dietary potassium. Deuterium substitution, as seen in Aldosterone-d4, has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs. It can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Deuterium labeled compound.
Catalog No. 50-004-36071 Supplier Medchemexpress LLC Supplier No. HY113313S2100UG
Aldosterone-d4 is a deuterium-labeled version of Aldosterone, which is the primary mineralocorticoid. This steroid hormone is synthesized and secreted in response to the activation of the renin-angiotensin system (RAS) or high dietary potassium. Deuterium substitution, as seen in Aldosterone-d4, has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs. It can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Deuterium labeled compound.