Learn More
Medchemexpress LLC Pregn-4-ene-3,20-dione-1,2-d2, 11,17,21-trihydroxy-, (11β)- | 1257650-73-9 | 364.47 | 5 MG
SDP
Catalog No. 5000388561
Encompass_Preferred
Hydrocortisone-d2 is a deuterium-labeled version of Hydrocortisone (Cortisol), a naturally occurring steroid hormone and glucocorticoid secreted by the adrenal cortex. This compound is intended for research use only and serves as a valuable tool in scientific studies.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Offers potential to affect pharmacokinetic and metabolic profiles of drugs due to deuteration.
  • Used in studies investigating the impact of deuterium substitution on pharmaceutical pharmacokinetics.
  • Used in research on the differential effects of hydrocortisone on tight junction proteins and allergic T-cell responses.
  • Utilized in studies exploring hydrocortisone's role in preserving the vascular barrier.
Catalog No. 50-003-88561 Supplier Medchemexpress LLC Supplier No. HYN0583S55MG
Hydrocortisone-d2 is a deuterium-labeled version of Hydrocortisone (Cortisol), a naturally occurring steroid hormone and glucocorticoid secreted by the adrenal cortex. This compound is intended for research use only and serves as a valuable tool in scientific studies.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Offers potential to affect pharmacokinetic and metabolic profiles of drugs due to deuteration.
  • Used in studies investigating the impact of deuterium substitution on pharmaceutical pharmacokinetics.
  • Used in research on the differential effects of hydrocortisone on tight junction proteins and allergic T-cell responses.
  • Utilized in studies exploring hydrocortisone's role in preserving the vascular barrier.