Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pregn-4-ene-3,20-dione-d9 | 15775-74-3 | MFCD01317491 | 99.7% | 323.52 | C21H21D9O2 | 10 MG
SDP

Catalog No. 5000296056
Encompass_Preferred

Progesterone-d9 is a deuterium-labeled analogue of the endogenous steroid hormone progesterone, provided as a solid for research use. It is intended for use as a tracer and internal standard in quantitative analytical methods and pharmacokinetic or metabolic studies.

  • Deuterium-labeled tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Chemical formula C21H21D9O2.
  • Molecular weight 323.52 g/mol.
  • Supplier-listed purity ~99.7% (verify for critical use).
  • Appearance: white to light yellow solid.
  • Solubility: DMSO 50 mg/mL; may require sonication and warming.
  • Storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Typical pack size: 10 mg.

Catalog No. 50-002-96056 Supplier Medchemexpress LLC Supplier No. HYN0437S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Progesterone-d9 is a deuterium-labeled analogue of the endogenous steroid hormone progesterone, provided as a solid for research use. It is intended for use as a tracer and internal standard in quantitative analytical methods and pharmacokinetic or metabolic studies.

  • Deuterium-labeled tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Chemical formula C21H21D9O2.
  • Molecular weight 323.52 g/mol.
  • Supplier-listed purity ~99.7% (verify for critical use).
  • Appearance: white to light yellow solid.
  • Solubility: DMSO 50 mg/mL; may require sonication and warming.
  • Storage: powder at -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Typical pack size: 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.