Learn More
Medchemexpress LLC Pregn-5-en-3β-ol | 2862-58-0 | 97.0% | 302.49 | 10 MG
SDP
Catalog No. 5000365578
Encompass_Preferred
Pregn-5-en-3β-ol is a steroid compound. When formulated as biscarboxylic acid hemiesters along with androst-5-en-3β-ol, it acts as a potent positive allosteric modulator of NMDA (N-methyl-D-aspartate) receptors, making it relevant for research into psychiatric disorders.
  • Applicable for research into psychiatric disorders
  • Solid, white to off-white in color
  • Ships at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • For research use only
Catalog No. 50-003-65578 Supplier Medchemexpress LLC Supplier No. 10MGHYW882667
Pregn-5-en-3β-ol is a steroid compound. When formulated as biscarboxylic acid hemiesters along with androst-5-en-3β-ol, it acts as a potent positive allosteric modulator of NMDA (N-methyl-D-aspartate) receptors, making it relevant for research into psychiatric disorders.
  • Applicable for research into psychiatric disorders
  • Solid, white to off-white in color
  • Ships at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • For research use only