Learn More
Medchemexpress LLC Pregn-5-en-3β-ol | 2862-58-0 | 97.0% | 302.49 | 25 MG
SDP
Catalog No. 5000365701
Encompass_Preferred
Pregn-5-en-3β-ol is a steroid compound that acts as a potent positive allosteric modulator of NMDA (N-methyl-D-aspartate) receptors when formulated as biscarboxylic acid hemiesters with androst-5-en-3β-ol. This compound is applicable for research into psychiatric disorders.
  • Steroid compound
  • Acts as a potent positive allosteric modulator of NMDA receptors
  • Applicable for research into psychiatric disorders
Catalog No. 50-003-65701 Supplier Medchemexpress LLC Supplier No. 25MGHYW882667
Pregn-5-en-3β-ol is a steroid compound that acts as a potent positive allosteric modulator of NMDA (N-methyl-D-aspartate) receptors when formulated as biscarboxylic acid hemiesters with androst-5-en-3β-ol. This compound is applicable for research into psychiatric disorders.
  • Steroid compound
  • Acts as a potent positive allosteric modulator of NMDA receptors
  • Applicable for research into psychiatric disorders