Learn More
Medchemexpress LLC Pregn-5-en-3β-ol | 2862-58-0 | 97.0% | 302.49 | 5 MG
SDP
Catalog No. 5000393451
Encompass_Preferred
Pregn-5-en-3β-ol is a steroid compound that functions as a potent positive allosteric modulator of NMDA (N-methyl-D-aspartate) receptors. It is relevant for research concerning psychiatric disorders.
  • Steroid compound
  • Functions as a potent positive allosteric modulator of NMDA receptors
  • Applicable for research into psychiatric disorders
  • For research use only
Catalog No. 50-003-93451 Supplier Medchemexpress LLC Supplier No. 5MGHYW882667
Pregn-5-en-3β-ol is a steroid compound that functions as a potent positive allosteric modulator of NMDA (N-methyl-D-aspartate) receptors. It is relevant for research concerning psychiatric disorders.
  • Steroid compound
  • Functions as a potent positive allosteric modulator of NMDA receptors
  • Applicable for research into psychiatric disorders
  • For research use only