Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-fluoro-11-hydroxy-16,17-(1-methylethylidene) | 3870-07-3 | MFCD09954497 | 99.4% | 476.53 g/mol | C26H33FO7 | 250 MG
SDP

Catalog No. 5000282346
Encompass_Preferred

Triamcinolone acetonide 21-acetate is a research-grade synthetic corticosteroid derivative supplied as a white to off-white solid. It is provided for laboratory and analytical applications as a biochemical reagent or reference standard and is not intended for human or clinical use.

  • High purity of 99.4%.
  • Solid, white to off-white powder.
  • Chemical formula C26H33FO7 and molecular weight 476.53 g/mol.
  • CAS number 3870-07-3 for unambiguous identification.
  • Storage: powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Suitable as an analytical standard or biochemical reagent in life-science research.

Catalog No. 50-002-82346 Supplier Medchemexpress LLC Supplier No. HYW340872250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triamcinolone acetonide 21-acetate is a research-grade synthetic corticosteroid derivative supplied as a white to off-white solid. It is provided for laboratory and analytical applications as a biochemical reagent or reference standard and is not intended for human or clinical use.

  • High purity of 99.4%.
  • Solid, white to off-white powder.
  • Chemical formula C26H33FO7 and molecular weight 476.53 g/mol.
  • CAS number 3870-07-3 for unambiguous identification.
  • Storage: powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Suitable as an analytical standard or biochemical reagent in life-science research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.