Learn More
Medchemexpress LLC Pregnan-20-one, 3,17,21-trihydroxy-, (3α,5β)- | 68-60-0 | MFCD00069490 | 98.2% | 350.49 g·mol⁻¹ | C21H34O4 | 5 MG
SDP
Catalog No. 5000264256
Encompass_Preferred
Tetrahydro-11-deoxycortisol is an endogenous urinary steroid metabolite supplied as a research reference standard for studies of 11β-hydroxylase deficiency and steroid metabolism. It is provided as a white to off-white solid suitable for analytical and biochemical applications.
  • High purity (>98.2% by HPLC).
  • Suitable as a reference standard for steroid metabolism and diagnostic research.
  • Solid form for accurate weighing and analytical preparation.
  • Molecular formula C21H34O4; molecular weight 350.49 g·mol⁻¹.
  • Available in small-quantity vials for method development and validation.
Catalog No. 50-002-64256 Supplier Medchemexpress LLC Supplier No. HY1134235MG
Tetrahydro-11-deoxycortisol is an endogenous urinary steroid metabolite supplied as a research reference standard for studies of 11β-hydroxylase deficiency and steroid metabolism. It is provided as a white to off-white solid suitable for analytical and biochemical applications.
  • High purity (>98.2% by HPLC).
  • Suitable as a reference standard for steroid metabolism and diagnostic research.
  • Solid form for accurate weighing and analytical preparation.
  • Molecular formula C21H34O4; molecular weight 350.49 g·mol⁻¹.
  • Available in small-quantity vials for method development and validation.