Learn More
Cliawaived Inc CLIAwaived,Inc. Urine Pregnancy Test Cassettes
SDP
Catalog No. NC1112619
Encompass_Preferred
(50/Pack) The all new CLIAwaived, Inc. Pregnancy Test (Cassette) is a qualitative immunoassay for the detection of human chorionic gonadatropin (hCG) in human urine for the early detection of pregnancy. hCG is a hormone produced by the placenta, which, in normal subjects, provides an early indication of pregnancy. Early testing and detection of hCG is vital for the health of mother and child, and to deliver reliable diagnostic information on which health care professionals can base optimal patient management strategies.
Catalog No. NC1112619 Supplier Cliawaived Inc Supplier No. CLIA022479
(50/Pack) The all new CLIAwaived, Inc. Pregnancy Test (Cassette) is a qualitative immunoassay for the detection of human chorionic gonadatropin (hCG) in human urine for the early detection of pregnancy. hCG is a hormone produced by the placenta, which, in normal subjects, provides an early indication of pregnancy. Early testing and detection of hCG is vital for the health of mother and child, and to deliver reliable diagnostic information on which health care professionals can base optimal patient management strategies.