Learn More
Cliawaived Inc CLIAwaived, Inc. Pregnancy Tests
SDP
Catalog No. NC1859167
Encompass_Preferred
(25 Tests)The CLIAwaived, Inc. Pregnancy Test (Cassette), 7mm hCG Top, is a rapid one step visual test for the qualitative detection of human chorionic gonadotropin hCG at 25mIU/mL in urine. FDA Cleared and CLIA Waived. (FDA 510K#: K984076)
This item is not returnable. View return policy
Catalog No. NC1859167 Supplier Cliawaived Inc Supplier No. CLIA022478
(25 Tests)The CLIAwaived, Inc. Pregnancy Test (Cassette), 7mm hCG Top, is a rapid one step visual test for the qualitative detection of human chorionic gonadotropin hCG at 25mIU/mL in urine. FDA Cleared and CLIA Waived. (FDA 510K#: K984076)