Learn More
Cliawaived Inc CLIAwaived, Inc. Pregnancy Urine Dip-Strips
SDP
Catalog No. NC1859166
Encompass_Preferred
(50 Tests)The CLIAwaived, Inc. Pregnancy Urine (Dip-Strip) test is a qualitative immunoassay for the detection of human chorionic gonadotropin (hCG) in human urine for the early detection of pregnancy. hCG is a hormone produced by the placenta, which, in normal subjects, provides an early indication of pregnancy. The pregnancy urine dip strip test provides a qualitative screening result and is for healthcare professional use only and not for self-testing. A more specific alternate chemical method must be used in order to obtain a confirmed analytical result.FDA Cleared and CLIA Waived. (FDA 510K#: K984079)
This item is not returnable. View return policy
Catalog No. NC1859166 Supplier Cliawaived Inc Supplier No. CLIA022471
(50 Tests)The CLIAwaived, Inc. Pregnancy Urine (Dip-Strip) test is a qualitative immunoassay for the detection of human chorionic gonadotropin (hCG) in human urine for the early detection of pregnancy. hCG is a hormone produced by the placenta, which, in normal subjects, provides an early indication of pregnancy. The pregnancy urine dip strip test provides a qualitative screening result and is for healthcare professional use only and not for self-testing. A more specific alternate chemical method must be used in order to obtain a confirmed analytical result.FDA Cleared and CLIA Waived. (FDA 510K#: K984079)