Learn More
Medchemexpress LLC Pregnanediol 3-glucuronide | 1852-49-9 | 98.0% | 496.63 g·mol⁻¹ | C27H44O8 | 5 MG
SDP
Catalog No. 5000299837
Encompass_Preferred
Pregnanediol 3-glucuronide is the primary urinary metabolite of the steroid hormone progesterone, formed by hepatic hydroxylation and glucuronidation. It is highly hydrophilic and readily excreted in urine, and is used in research and clinical assays as a biochemical marker to monitor ovulation, evaluate luteal function, and study reproductive endocrinology.
  • Primary urinary metabolite of progesterone.
  • Highly hydrophilic and readily excreted in urine.
  • Serves as a biochemical marker to monitor ovulation and luteal function.
  • Suitable for use in analytical and clinical assays.
  • Available in small, research-grade quantities for reference standards.
Catalog No. 50-002-99837 Supplier Medchemexpress LLC Supplier No. HYW0400475MG
Pregnanediol 3-glucuronide is the primary urinary metabolite of the steroid hormone progesterone, formed by hepatic hydroxylation and glucuronidation. It is highly hydrophilic and readily excreted in urine, and is used in research and clinical assays as a biochemical marker to monitor ovulation, evaluate luteal function, and study reproductive endocrinology.
  • Primary urinary metabolite of progesterone.
  • Highly hydrophilic and readily excreted in urine.
  • Serves as a biochemical marker to monitor ovulation and luteal function.
  • Suitable for use in analytical and clinical assays.
  • Available in small, research-grade quantities for reference standards.