Learn More
Medchemexpress LLC Pregnanediol-d5 | 00-00-0 | 99.1% | 325.54 g/mol | C21H31D5O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY107850S1MG
Encompass_Preferred
Pregnanediol-d5 is the deuterium-labeled form of pregnanediol supplied as a powder for use as a stable isotope internal standard in quantitative analytical methods. It is intended for use as a tracer or internal standard in NMR, GC-MS, and LC-MS workflows, and is stable under recommended cold storage conditions.
  • Deuterated internal standard for quantitative analysis.
  • High chemical purity and isotopic enrichment.
  • Available in small milligram quantities for analytical workflows.
  • Stable as a powder under recommended cold storage conditions.
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
Catalog No. 50-002-42277