Learn More
Medchemexpress LLC Pregnanetriol-d4 | 2687960-83-2 | 99.84% | C21H32D4O3 | 1 MG
SDP
Catalog No. 5000353656
Encompass_Preferred
Pregnanetriol-d4 is a deuterium-labeled form of Pregnanetriol, suitable for research applications. It can serve as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuterium substitution, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has attracted attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Available as a white to off-white solid
  • Intended for research use only
Catalog No. 50-003-53656 Supplier Medchemexpress LLC Supplier No. HY142567S1MG
Pregnanetriol-d4 is a deuterium-labeled form of Pregnanetriol, suitable for research applications. It can serve as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuterium substitution, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has attracted attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Available as a white to off-white solid
  • Intended for research use only