Learn More
Medchemexpress LLC Pregnanolone sulfate pyridinium | 124107-39-7 | 99.8% | 477.66 | C26H39NO5S | 100mg
SDP
Catalog No. 5000269293
Encompass_Preferred
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]
Catalog No. 50-002-69293 Supplier Medchemexpress LLC Supplier No. HYW742957100MG
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]