Learn More
Medchemexpress LLC Pregnanolone sulfate (pyridinium) | 124107-39-7 | 99.8% | 477.66 | C26H39NO5S | 10mg
SDP
Catalog No. 5000269294
Encompass_Preferred
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]
Catalog No. 50-002-69294 Supplier Medchemexpress LLC Supplier No. HYW74295710MG
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]