Learn More
Medchemexpress LLC Pregnanolone sulfate (pyridinium) | 124107-39-7 | Not provided | 99.8% | 477.66 | C26H39NO5S | 25mg
SDP
Catalog No. 5000270499
Encompass_Preferred
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]
Catalog No. 50-002-70499 Supplier Medchemexpress LLC Supplier No. HYW74295725MG
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]