Learn More
Cayman Chemical PregndIol-3-GlucuronIdeP 96Wel
SDP
Catalog No. 502895157
Encompass_Preferred
Cayman’s Pregnanediol-3-Glucuronide (PDG) ELISA Kit is a competitive assay that can be used for quantification of pregnanediol metabolites in urine. The assay has a range from 0.4-50 ng/ml and a sensitivity (80% B/B0) of approximately 0.94 ng/ml.
Catalog No. 50-289-5157 Supplier Cayman Chemical Supplier No. 50130096 WELL
Cayman’s Pregnanediol-3-Glucuronide (PDG) ELISA Kit is a competitive assay that can be used for quantification of pregnanediol metabolites in urine. The assay has a range from 0.4-50 ng/ml and a sensitivity (80% B/B0) of approximately 0.94 ng/ml.