Learn More
Medchemexpress LLC Pregnenolone (3β-Hydroxy-5-pregnen-20-one) | 145-13-1 | >99.9% | 25 G
SDP
Catalog No. 5000407436
Encompass_Preferred
Pregnenolone (3β-Hydroxy-5-pregnen-20-one) is a potent neurosteroid and the primary precursor for various steroid hormones, including steroid ketones. It functions as a signaling-specific inhibitor of the cannabinoid CB1 receptor, thereby inhibiting the effects of tetrahydrocannabinol (THC) that are mediated by CB1 receptors. Pregnenolone can protect the brain from cannabis intoxication. Additionally, it acts as a TRPM3 channel activator and can weakly activate TRPM1 channels.
  • Potent neurosteroid
  • Primary precursor for various steroid hormones
  • Signaling-specific inhibitor of CB1 receptor
  • Inhibits THC effects mediated by CB1 receptors
  • Protects the brain from cannabis intoxication
  • Acts as a TRPM3 channel activator
  • Weakly activates TRPM1 channels
Catalog No. 50-004-07436 Supplier Medchemexpress LLC Supplier No. 25GHYB0151
Pregnenolone (3β-Hydroxy-5-pregnen-20-one) is a potent neurosteroid and the primary precursor for various steroid hormones, including steroid ketones. It functions as a signaling-specific inhibitor of the cannabinoid CB1 receptor, thereby inhibiting the effects of tetrahydrocannabinol (THC) that are mediated by CB1 receptors. Pregnenolone can protect the brain from cannabis intoxication. Additionally, it acts as a TRPM3 channel activator and can weakly activate TRPM1 channels.
  • Potent neurosteroid
  • Primary precursor for various steroid hormones
  • Signaling-specific inhibitor of CB1 receptor
  • Inhibits THC effects mediated by CB1 receptors
  • Protects the brain from cannabis intoxication
  • Acts as a TRPM3 channel activator
  • Weakly activates TRPM1 channels