Learn More
Apexbio Technology LLC Pregnenolone 145-13-1 5g
SDP
Catalog No. 502240656
Encompass_Preferred
Pregnenolone (CAS 145-13-1) also known as 3 -hydroxypregn-5-en-20-one or P5 is an endogenous neurosteroid and precursor of nearly all other steroid hormones including DHEA testosterone progesterone estrogens and cortisol In the central nervous system pregnenolone modulates synaptic transmission and has been shown to influence learning and memory processes In rodent models its sulfate derivative (PREGS) improves age-related impairments in memory retention in a dose-dependent manner Clinical findings on its cognitive benefits are mixed with some studies reporting enhanced memory in individuals with low baseline PREGS levels Pregnenolone is widely used in neuroscience research on neurosteroid function and cognitive aging
This item is not returnable. View return policy
Catalog No. 50-224-0656 Supplier Apexbio Technology LLC Supplier No. B15125000
Pregnenolone (CAS 145-13-1) also known as 3 -hydroxypregn-5-en-20-one or P5 is an endogenous neurosteroid and precursor of nearly all other steroid hormones including DHEA testosterone progesterone estrogens and cortisol In the central nervous system pregnenolone modulates synaptic transmission and has been shown to influence learning and memory processes In rodent models its sulfate derivative (PREGS) improves age-related impairments in memory retention in a dose-dependent manner Clinical findings on its cognitive benefits are mixed with some studies reporting enhanced memory in individuals with low baseline PREGS levels Pregnenolone is widely used in neuroscience research on neurosteroid function and cognitive aging