Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pregnenolone, 98%, Spectrum™ Chemical
SDP

Catalog No. 18612947 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 kg
25 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-947 5 kg Amber Glass Bottle
18-612-945 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-612-947 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14225KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145-13-1

Specifications

CAS Max % 1
Boiling Point 443.26°C (829.87°F)
Loss on Drying 0.005
Melting Range 183° to 194°C
Packaging Amber Glass Bottle
Quantity 5 kg
Specific Rotation +27° to +30°
Molecular Weight (g/mol) 316.47
Percent Purity 0.98
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Pregnenolone
Show More Show Less

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.