Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pregnenolone, 99%, Thermo Scientific Chemicals

Catalog No. AC164421000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164421000 100 g Glass bottle
AC164420050 5 g Glass bottle
AC164420250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164421000 Supplier Thermo Scientific Chemicals Supplier No. 164421000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-13-1
Molecular Formula C21H32O2
Molecular Weight (g/mol) 316.47
MDL Number MFCD00003628
InChI Key ORNBQBCIOKFOEO-OYZZJKLHSA-N
Synonym 3-hydroxypregn-5-en-20-one
PubChem CID 44307923
IUPAC Name 1-[(10R,13S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMILES CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 190°C to 193°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 7851
Specific Rotation Condition +28.50° (20°C c=1,EtOH)
Specific Rotation + 28.5
Solubility Information Solubility in water: slightly soluble. Other solubilities: solubility (g/100 mL of solution):,carbon terachloride 0.5 / petroleum ether 0.1 / et,acetone 0.6 / chloroform 17.0 / ethanol 1.9 / benz,isopropanol 1.5,solubility (g/100 mL of solvent):
Formula Weight 316.47
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Pregnenolone
Show More Show Less
EINECSNumber 205-647-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.