Learn More
Medchemexpress LLC Pregnenolone acetate | 1778-02-5 | 98.4% | 1 ML
SDP
Catalog No. 5000411453
Encompass_Preferred
Pregnenolone acetate is a nonmetabolized Pregnenolone analog that promotes neurite extension and induces changes in growth cone morphology in primary cultures of cerebellar granule neurons. It is a neurosteroid that increases microtubule polymerization and shows potential for neurodevelopmental diseases research.
  • Promotes neurite extension
  • Induces changes in growth cone morphology
  • Increases microtubule polymerization
  • Potential for neurodevelopmental diseases research
Catalog No. 50-004-11453 Supplier Medchemexpress LLC Supplier No. HYB174010MM/1ML
Pregnenolone acetate is a nonmetabolized Pregnenolone analog that promotes neurite extension and induces changes in growth cone morphology in primary cultures of cerebellar granule neurons. It is a neurosteroid that increases microtubule polymerization and shows potential for neurodevelopmental diseases research.
  • Promotes neurite extension
  • Induces changes in growth cone morphology
  • Increases microtubule polymerization
  • Potential for neurodevelopmental diseases research