Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pregnenolone Acetate, MP Biomedicals™

Catalog No. ICN10272001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10272001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10272001 Supplier MP Biomedicals, Inc Supplier No. 0210272001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1778-02-5
Molecular Formula C23H34O3
Molecular Weight (g/mol) 358.522
InChI Key CRRKVZVYZQXICQ-BODHGXFRSA-N
Synonym 1s,5s,10s,11s,14s,15s,2r-14-acetyl-2,15-dimethyltetracyclo 8.7.0.0<2,7>.0<11 ,15> heptadec-7-en-5-yl acetate, 11as-1-acetyl-9a,11a-dimethyl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl acetate, 13s-17-ethanoyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ethanoate, acetic acid 13s-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ester
PubChem CID 18530247
IUPAC Name [(13S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C

Specifications

Quantity 1 g
Formula Weight 358.5
Physical Form Solid
Chemical Name or Material Pregnenolone Acetate
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.