Learn More
Medchemexpress LLC Pregnenolone monosulfate | 1247-64-9 | 99.96% | 1 ML
SDP
Catalog No. 5000412696
Encompass_Preferred
Pregnenolone monosulfate is a powerful neurosteroid and the main precursor of various steroid hormones, including steroid ketones. It functions as a signaling-specific inhibitor of the cannabinoid CB1 receptor, inhibiting the effects of tetrahydrocannabinol (THC) that are mediated by CB1 receptors. It can protect the brain from cannabis intoxication and also acts as a TRPM3 channel activator and can weakly activate TRPM1 channels.
  • Powerful neurosteroid
  • Main precursor of various steroid hormones
  • Signaling-specific inhibitor of cannabinoid CB1 receptor
  • Inhibits THC effects mediated by CB1 receptors
  • Protects the brain from cannabis intoxication
  • TRPM3 channel activator
  • Weakly activates TRPM1 channels
  • For research use only
  • Solid appearance, white to off-white color
  • Initial source: Endogenous metabolite
  • Structure classification: Steroids
Catalog No. 50-004-12696 Supplier Medchemexpress LLC Supplier No. HYB173910MM/1ML
Pregnenolone monosulfate is a powerful neurosteroid and the main precursor of various steroid hormones, including steroid ketones. It functions as a signaling-specific inhibitor of the cannabinoid CB1 receptor, inhibiting the effects of tetrahydrocannabinol (THC) that are mediated by CB1 receptors. It can protect the brain from cannabis intoxication and also acts as a TRPM3 channel activator and can weakly activate TRPM1 channels.
  • Powerful neurosteroid
  • Main precursor of various steroid hormones
  • Signaling-specific inhibitor of cannabinoid CB1 receptor
  • Inhibits THC effects mediated by CB1 receptors
  • Protects the brain from cannabis intoxication
  • TRPM3 channel activator
  • Weakly activates TRPM1 channels
  • For research use only
  • Solid appearance, white to off-white color
  • Initial source: Endogenous metabolite
  • Structure classification: Steroids