Learn More
TARGETMOL CHEMICALS INC Pregnenolone monosulfate 25MG
SDP
Catalog No. 502877497
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Pregnenolone monosulfate (3beta-Hydroxy-5-pregnen-20-one monosulfate) is a potent nootropic steroid that positively regulates NMDA recordings and is utilized in the study of neuroticism and depression. Purity 99.46%
Catalog No. 50-287-7497 Supplier TARGETMOL CHEMICALS INC Supplier No. T1253625MG
Also available in 1 mL, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Pregnenolone monosulfate (3beta-Hydroxy-5-pregnen-20-one monosulfate) is a potent nootropic steroid that positively regulates NMDA recordings and is utilized in the study of neuroticism and depression. Purity 99.46%