Learn More
Cayman Chemical Pregnenolone Carbonitrile, Purity: ≥97%, CAS: 1434-54-4, 100 mg
SDP
Catalog No. 502973304
Encompass_Preferred
A catatoxic steroid that acts as an agonist of the rodent PXR at µM concentrations; mediates induction of hepatic CYP3A, Oatp2, and Cyp7a1, regulating both xenobiotic and bile acid homeostasis
Catalog No. 50-297-3304 Supplier Cayman Chemical Supplier No. 16343100 MG
A catatoxic steroid that acts as an agonist of the rodent PXR at µM concentrations; mediates induction of hepatic CYP3A, Oatp2, and Cyp7a1, regulating both xenobiotic and bile acid homeostasis