Learn More
Cayman Chemical Pregnolon sulfatesodIum 50mg
SDP
Catalog No. 502983795
Encompass_Preferred
A metabolite of pregnenolone that modulates NMDA receptor responses to exogenously applied glutamate and stimulates TRPM3
Catalog No. 50-298-3795 Supplier Cayman Chemical Supplier No. 2100450 MG
A metabolite of pregnenolone that modulates NMDA receptor responses to exogenously applied glutamate and stimulates TRPM3