Learn More
Cayman Chemical Pregnanetriol, Purity: ≥95%, CAS: 1098-45-9, 10 mg
SDP
Catalog No. 502948867
Encompass_Preferred
A metabolite of 17α-hydroxyprogesterone; urinary levels are elevated in patients with 21-hydroxylase deficiency and congenital adrenal hyperplasia
Catalog No. 50-294-8867 Supplier Cayman Chemical Supplier No. 3331210 MG
A metabolite of 17α-hydroxyprogesterone; urinary levels are elevated in patients with 21-hydroxylase deficiency and congenital adrenal hyperplasia