Learn More
Waters Corp Preheater Tubing Kit, with Thermal Gaskets, Left
Catalog No. 502391338 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides replacement tubing assemblies and associated thermal gaskets for the preheater of a Left Inlet ACQUITY UPLC Column Manager.
  • Spare parts kit
  • System type: UPC2
  • Replacement schedule: As required
Catalog No. 50-239-1338 Supplier Waters Corp Supplier No. 700003246
This kit provides replacement tubing assemblies and associated thermal gaskets for the preheater of a Left Inlet ACQUITY UPLC Column Manager.
  • Spare parts kit
  • System type: UPC2
  • Replacement schedule: As required