Learn More
Active Motif Preindex Tn5 Transposomes 1x96
SDP
Catalog No. NC2586645
Encompass
Pre-indexed Assembled Tn5 Transposomes are provided in a flexible breakaway 96-well plate format to enable higher throughput ATAC-Seq assays. Each well contains a transposome loaded with a unique i5 and i7 index, enabling direct indexing during tagmentation. By having a unique i7 & i5 Indexed Primer combination in each well, it is easy to perform multiple ATAC-Seq reactions without having to pipette primers separately and keep track of the combinations.
Catalog No. NC2586645 Supplier Active Motif Supplier No. 53152
Pre-indexed Assembled Tn5 Transposomes are provided in a flexible breakaway 96-well plate format to enable higher throughput ATAC-Seq assays. Each well contains a transposome loaded with a unique i5 and i7 index, enabling direct indexing during tagmentation. By having a unique i7 & i5 Indexed Primer combination in each well, it is easy to perform multiple ATAC-Seq reactions without having to pipette primers separately and keep track of the combinations.