Learn More
Apexbio Technology LLC Preladenant 377727-87-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062836
Encompass_Preferred
Preladenant is a potent and selective antagonist targeting the adenosine A2A receptor (Ki 1 1 nM) developed from structural modifications of SCH 58261 Structurally it is a methoxy derivative carrying enhanced affinity and selectivity towards human and rat A2A receptors over A1 A2B and A3 subtypes Mechanistically Preladenant competitively blocks A2A receptor-mediated adenylate cyclase activation In preclinical studies Preladenant reduces Parkinsonian symptoms in primate and rodent models alone or in combination with L-Dopa thus widely employed in experimental research for Parkinson s disease and associated movement disorders
Catalog No. 50-000-62836 Supplier Apexbio Technology LLC Supplier No. A37355.1
Preladenant is a potent and selective antagonist targeting the adenosine A2A receptor (Ki 1 1 nM) developed from structural modifications of SCH 58261 Structurally it is a methoxy derivative carrying enhanced affinity and selectivity towards human and rat A2A receptors over A1 A2B and A3 subtypes Mechanistically Preladenant competitively blocks A2A receptor-mediated adenylate cyclase activation In preclinical studies Preladenant reduces Parkinsonian symptoms in primate and rodent models alone or in combination with L-Dopa thus widely employed in experimental research for Parkinson s disease and associated movement disorders