Learn More
Sino Biological PRELP / Prolargin Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 13538T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PRELP / Prolargin (rh PRELP / Prolargin; Catalog#13538-HNCH; NP_002716.1; Gln21-Ile382). PRELP / Prolargin specific IgG was purified by Human PRELP / Prolargin affinity chromatography.
Catalog No. 50-224-8711