Learn More
Cole-Parmer Cole-Parmer Stuart Premium Digital Stirring Hot Plate, Ceramic, Blue; 120 VAC
Catalog No. 501960401 Shop All Cole-Parmer Products
Encompass_Preferred
These custom designed stirring hot plates provide accurate stirring and temperature control. They feature a hot surface warning light that flashes when the plate temperature is above 50C. Its durable front panel resists spills and chemicals. The plates are available in various interface, plate surface,and body color configurations.
Catalog No. 50-196-0401 Supplier Cole-Parmer Supplier No. 0480680
These custom designed stirring hot plates provide accurate stirring and temperature control. They feature a hot surface warning light that flashes when the plate temperature is above 50C. Its durable front panel resists spills and chemicals. The plates are available in various interface, plate surface,and body color configurations.