Learn More
MSC Smith Performance Tank Sprayer, 3 Gal, Chemical Safe, 1 ea
Catalog No. 502866810
Encompass
This 3 gal. chemical safe hand sprayer is designed for various applications, including cleaning, degreasing, and applying lawn or pest control chemicals. It features a durable polyethylene tank and a reinforced hose for reliable performance.
  • Integrated splash cap directs vapors and liquids away when opened.
  • Highly efficient pump requires fewer strokes for pressurization.
  • Buna gaskets and seals provide chemical protection for water-based applications.
  • Durable 18 in aluminum wand and 36 in reinforced hose with comfort grip shut-off.
  • Adjustable brass nozzle offers stream to cone spray patterns.
Catalog No. 50-286-6810 Supplier MSC Supplier No. 30302319
This 3 gal. chemical safe hand sprayer is designed for various applications, including cleaning, degreasing, and applying lawn or pest control chemicals. It features a durable polyethylene tank and a reinforced hose for reliable performance.
  • Integrated splash cap directs vapors and liquids away when opened.
  • Highly efficient pump requires fewer strokes for pressurization.
  • Buna gaskets and seals provide chemical protection for water-based applications.
  • Durable 18 in aluminum wand and 36 in reinforced hose with comfort grip shut-off.
  • Adjustable brass nozzle offers stream to cone spray patterns.