Learn More
System Biosciences LLC pMC.CMV-GFP-SV40PolyA Positive Control MinicirclePre-made Minicircle CMV-GFP-SV40PolyA (positive control) = 40 µg
SDP
Catalog No. NC2042718
Encompass_Preferred
Easily implement positive controls for your minicircle projects with pMC.CMV-GFP-SV40PolyA, a ready-to-transfect, pre-made, positive control minicircle. Maximize safety with Minicircle Technology—episomal expression free from foreign DNA Jump start your minicircle positive controls and troubleshooting with the ready-to-transfect pMC.CMV-GFP-SV40PolyA Pre-made Positive Control Minicircle (Cat.# MN601MC-1)., you can easily implement positive controls or troubleshoot your minicircle projects. This plasmid uses the strong CMV promoter to drive GFP expression so you can track transfection efficiencies and more with GFP imaging.
Catalog No. NC2042718 Supplier System Biosciences LLC Supplier No. MN601MC1
Easily implement positive controls for your minicircle projects with pMC.CMV-GFP-SV40PolyA, a ready-to-transfect, pre-made, positive control minicircle. Maximize safety with Minicircle Technology—episomal expression free from foreign DNA Jump start your minicircle positive controls and troubleshooting with the ready-to-transfect pMC.CMV-GFP-SV40PolyA Pre-made Positive Control Minicircle (Cat.# MN601MC-1)., you can easily implement positive controls or troubleshoot your minicircle projects. This plasmid uses the strong CMV promoter to drive GFP expression so you can track transfection efficiencies and more with GFP imaging.