Learn More
Surgical Design Premier Adjustable Surgical Knife Scalpel Handle
Catalog No. 22079772 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 3
Catalog No. Size
22-079-772 # 3
1 options
Catalog No. 22-079-772 Supplier Surgical Design Supplier No. 270A
Add to Cart
Add to Cart
Surgical Design™ Premier Adjustable Surgical Knife Scalpel Handle used for sharp and precise dissection that features a unique multi-angle configuration. The knife handle comes with patterned surface for enhanced comfort and secure non-slip grip.
  • Surgical Design™ Premier Adjustable Surgical Knife Scalpel Handle is a uniquely designed to provide the user with multiple, adjustable angle configurations than the standard scalpel. This All-In-One scalpel reduces the need of multiple scalpels which decreases the rate of injury and improves efficiency during experimental and medical procedures.
    • Surgical Design™ Premier line exceeds performance and quality expectations. Guaranteed.
    • Unique adjustable pivot – Six angle configuration (Angles range up to 180°)
    • Handle designed and engineered for precision work
    • Comfortable and resuable
    • Blades slide on smoothly and securely
    • Superb quality and workmanship
    • Premium surgical grade stainless steel
    • Corrosion protected by passivation and ultrasonic cleaning
    • Sterilizable by Etoh/gamma radiation
    • Autoclavable
Specifications
Blade Material Stainless Steel
Product Type Scalpel Handle
Size # 3
Length (English) 5.75 in.
Length (Metric) 14.61cm
For Use With (Equipment) Compatible with 10 Series Disposable Scalpel Blade (Sizes # 10 to # 15)
Description Satin, Anti-glare, Adjustable Scalpel
Disposable Reusable
Autoclavable Autoclavable