Learn More
Leica Biosystems PREMIER COVERGLASS 24X50 CASE
Catalog No. NC0106068
Encompass_Preferred
Premier Cover Glass has been specially designed to work with automated coverslippers. Extra processes to prevent sticking are employed during manufacturing. Premier coverglass available in multiple sizes. Thinness: 013mm - 0.17mm Sizes range from 22 X 40mm to 24 x 60mm as noted below.
Catalog No. NC0106068 Supplier Leica Biosystems Supplier No. 3800145ACS
Premier Cover Glass has been specially designed to work with automated coverslippers. Extra processes to prevent sticking are employed during manufacturing. Premier coverglass available in multiple sizes. Thinness: 013mm - 0.17mm Sizes range from 22 X 40mm to 24 x 60mm as noted below.