Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Premier™ Work Boots

Catalog No. 19034462D Shop All Tingley Rubber Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-034-462D 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-034-462D Supplier Tingley™ Supplier No. 93245.09
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide excellent slip performance on surfaces

Made from high-molecular-weight PVC resins and specialty plasticizers that combine to provide protection from concentrates of fats, certain acids, hydrocarbons, caustics, and other chemicals. Work boots are suitable for food processing, particularly red meat, turkey, dairy and beverage processing; also ideal for chemical and petroleum industrial applications.

  • Reinforced molded shank supports arch, eliminating the need for steel
  • Cushioned insole absorbs three times its weight in perspiration
  • Flexible upper for comfort and mobility
  • Durable outsole with Chevron Plus™ tread for slip and abrasion resistance
  • Waterproof
  • 15 in. high (38cm)
  • Brick red upper with cream Chevron Plus outsole
  • Boots are EH rated

Compliance

Steel-toe meets ASTM™ F 2413-05 M/I/75/C/75.

Specifications

Certifications/Compliance ASTM
Color Red, Cream
For Use With (Application) Excellent choice for food processing and petrochemical applications
Height (English) 15 in.
Material PVC
Size 9
Sole Features Removable contour cushion insoles, Chevron-Plus outsole
Toe Material Steel
Waterproof Yes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.