Learn More
Premier Lab Supply Polypropylene 4.0μm (0.16mil) Continuous Roll – PN# TF-240
SDP
Catalog No. NC1598520
Encompass_Preferred
Polypropylene film 4.0μm (0.16mil) continuous roll with Dimensions: 3” x 300’ (76mm x 91.4m) (PN# TF-240). General purpose film with excellent X-ray transmission and chemical resistance, and lot number tracking system.
Catalog No. NC1598520 Supplier Premier Lab Supply Supplier No. TF240
Polypropylene film 4.0μm (0.16mil) continuous roll with Dimensions: 3” x 300’ (76mm x 91.4m) (PN# TF-240). General purpose film with excellent X-ray transmission and chemical resistance, and lot number tracking system.