Learn More
LabScientific Wax Trimmer, Digital Display
Catalog No. NC0310900
Encompass
Our Wax Trimmer has a compact and powerful build that is perfect for medical and research laboratories. It boasts a magnetically secured tray and a disposable tray liner that collects melted paraffin, making for easy and clean disposal. It is efficient and practical for removing excess paraffin from tissue cassettes. With this wax trimmer, there is no need to scrape the collection tray. Our convenient PE disposable tray liner allows easy disposal of the runoff wax. No need to scrape out the collection tray!Features: Multiple blocks may be trimmed simultaneously. Grooved surface for proper wax drainage. Two temperature settings low setting is approximately 80F and high setting is approximately 90F. UL listed power cord. Includes 5 disposable drip tray liners (additional liners available separately).
Catalog No. NC0310900 Supplier LabScientific Supplier No. XH90
Our Wax Trimmer has a compact and powerful build that is perfect for medical and research laboratories. It boasts a magnetically secured tray and a disposable tray liner that collects melted paraffin, making for easy and clean disposal. It is efficient and practical for removing excess paraffin from tissue cassettes. With this wax trimmer, there is no need to scrape the collection tray. Our convenient PE disposable tray liner allows easy disposal of the runoff wax. No need to scrape out the collection tray!Features: Multiple blocks may be trimmed simultaneously. Grooved surface for proper wax drainage. Two temperature settings low setting is approximately 80F and high setting is approximately 90F. UL listed power cord. Includes 5 disposable drip tray liners (additional liners available separately).