Learn More
SCP Science CONOSTAN PremiSolv, 1 Gallon, Zero-odor Alternative to Kerosene or Xylene
Catalog No. NC9776045
Encompass
This item has a minimum qty of 2 per supplier requirements.
PremiSolv is a zero-odor alternative to kerosene or xylene for use as a diluent or zero-point standard in ICP/DCP analysis of metals in oil and other organic fluids. PremiSolv Features: Extremely low odor - for a safer, more comfortable working environment Extremely low toxicity - compared with kerosene or xylene Extremely low metal content - comes with a certificate of analysis listing the concentrations of 33 different metals and sulfur Non-hazardous - for shipping and disposal
Catalog No. NC9776045 Supplier SCP Science Supplier No. 150700003
Add to Cart
Add to Cart
This item has a minimum qty of 2 per supplier requirements.
PremiSolv is a zero-odor alternative to kerosene or xylene for use as a diluent or zero-point standard in ICP/DCP analysis of metals in oil and other organic fluids. PremiSolv Features: Extremely low odor - for a safer, more comfortable working environment Extremely low toxicity - compared with kerosene or xylene Extremely low metal content - comes with a certificate of analysis listing the concentrations of 33 different metals and sulfur Non-hazardous - for shipping and disposal