Learn More
SCP Science CONOSTAN PremiSolv, 5 Gallons, Zero-odor Alternative to Kerosene or Xylene
Catalog No. NC9795865
Encompass
Conostan PremiSolv, 5 Gallons. PremiSolv ICP solvent is a replacement for low odor kerosene and xylene for use as a diluent in ICP/DCP analysis of metals in oil and other organic fluids. Features: Certificate of analysis listing the concentration of 34 different metals and sulphur;a(TM) extremely low toxicity compared to kerosene or xylene; no sulfur/aromatic odor for increased worker; comfort and less exposure; non-hazardous for shipping/disposal; dilution of samples and standards
Catalog No. NC9795865 Supplier SCP Science Supplier No. 150700002
Conostan PremiSolv, 5 Gallons. PremiSolv ICP solvent is a replacement for low odor kerosene and xylene for use as a diluent in ICP/DCP analysis of metals in oil and other organic fluids. Features: Certificate of analysis listing the concentration of 34 different metals and sulphur;a(TM) extremely low toxicity compared to kerosene or xylene; no sulfur/aromatic odor for increased worker; comfort and less exposure; non-hazardous for shipping/disposal; dilution of samples and standards