Learn More
Diagenode Inc PREMIUM BISULFITE KIT,50 RXNS
Supplier:  Diagenode Inc C02030030
Encompass
Diagenode's Premium Bisulfite Kit rapidly converts DNA through bisulfite treatment. Our conversion reagent is added directly to DNA, requires no intermediate steps, and results in high yields of DNA ready for downstream analysis methods including PCR and Next-Generation Sequencing.
Catalog No. NC1924929