Learn More
ML Kishigo Premium Black Series™ Surveyors Vest
Catalog No. 19441112
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-441-112 Lime Small
19-441-113 Lime Medium
19-441-122 Orange Large
19-441-124 Orange 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-441-112 Supplier ML Kishigo Supplier No. S5002S
Please call customer service for current pricing. 1-800-766-7000
This item is not returnable. View return policy
ML Kishigo Premium Black Series™ Surveyors Vest with ANSI 107 Class 2 Safety Compliance
  • Solid polyester front with mesh back
  • Heavy duty black bottom to help maintain a clean appearance
  • Zipper front closure reinforced with durable webbing
  • 2 in. wide high performance reflective material laminated to 3 in. contrasting color
  • Left and right mic tabs (Size small only has left mic tab)
  • 2 lapel grommets
  • Padded neck for comfort
Specifications
Construction Solid Front, Mesh Back
Certifications/Compliance ANSI 107 Class 2
Color Lime
Size Small
Material Polyester
Length (Metric) 70 cm