Learn More
Kishigo Premium Brilliant Series™ Heavy Duty Vest
Catalog No. 19220833
Click to view available options
Size:
Small
3X-Large
Color:
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-220-833 3X-Large Lime
19-220-838 Small Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-220-833 Supplier Kishigo Supplier No. 15103X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Created for maximum visibility. ML Kishigo™ Premium Brilliant Series™ Heavy Duty Vest enhances low light visibility and provides instant differentiation between worker and the work zone.
  • Made of Ultra-Cool™ 100% polyester mesh
  • Brilliant Trim reflective edging improves low light visibility
  • Zipper front closure reinforced with durable webbing
  • 2 in. wide high performance reflective material with 3 in. breathable contrasting color
  • Features left and right chest mic tabs
Pockets:
  • Two lower outside cargo pockets with grommets and adjustable flaps
  • Right chest 2-tier pencil pocket
  • Left chest radio pocket with flap
  • Two inside patch pockets
Compliance
Meets ANSI™ /ISEA 107-2015, Type R, Class 2 standards.
Specifications
Closure Type Zipper
Certifications/Compliance ANSI/ISEA 107-2015, Type R, Class 2
Color Lime
Size 3X-Large
Material Polyester Mesh
No. of Pockets 6
Tape Type 2 in. Reflective stripes